Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Candida Albicans Antibody (231/439), Novus Biologicals™
SDP

Catalog No. NB10062428 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10062428 0.1 mg
NB017473 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB10062428 Supplier Novus Biologicals Supplier No. NB10062428
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Candida Albicans Monoclonal specifically detects Candida Albicans in Yeast samples. It is validated for ELISA.

Specifications

Antigen Candida Albicans
Applications ELISA
Classification Monoclonal
Clone 231/439
Conjugate Unconjugated
Dilution ELISA 1:100-1:2000
Gene Alias C albicans, C. albicans, Thrush
Host Species Mouse
Immunogen Native
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This product recognizes Candida albicans, a yeast and opportunistic pathogen in humans.
Target Species Yeast
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgM
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.