Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Candida Albicans Antibody, Biotin, Novus Biologicals™
SDP

Catalog No. NB10065053 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10065053 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10065053 Supplier Novus Biologicals Supplier No. NB10065053
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Candida Albicans Polyclonal specifically detects Candida Albicans in Yeast samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry (Frozen).

Specifications

Antigen Candida Albicans
Applications ELISA, Immunohistochemistry, Immunohistochemistry (Frozen)
Classification Polyclonal
Concentration 4.0mg/mL
Conjugate Biotin
Dilution ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500
Formulation PBS (pH 7.2) with 0.05% Sodium Azide
Gene Alias C albicans, C. albicans, Thrush
Host Species Rabbit
Immunogen Candida albicans, type A
Purification Method IgG purified
Quantity 0.5 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity NB100-64750 reacts with numerous proteins in a soluble C. albicans extract in immunoelectrophoresis. Candida albicans is the most frequently isolated fungal pathogen of humans. It principally infects the skin and mucosal surfaces causing candidiasis or thrush. This antibody has not been absorbed and cross reacts with other yeasts.
Target Species Yeast
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.