Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Canine Distemper Virus Antibody (DV2-12), DyLight 550, Novus Biologicals™
SDP

Catalog No. NB10064816R Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.125 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10064816R 0.125 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10064816R Supplier Novus Biologicals Supplier No. NB10064816R
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Canine Distemper Virus Monoclonal specifically detects Canine Distemper Virus in Virus samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot.

Specifications

Antigen Canine Distemper Virus
Applications ELISA, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone DV2-12
Conjugate DyLight 550
Dilution Western Blot, ELISA, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias CDV
Host Species Mouse
Immunogen Made to Viral CANINE DISTEMPER VIRUS
Purification Method Protein A purified
Quantity 0.125 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity NB100-64816 recognizes canine distemper virus nucleoprotein. In Western blots the antibody recognizes bands of 76kD, 68kD, 54kD and 32kD under reducing conditions. This antibody is useful for the detection of CDV in immunofluorescence procedures, in which the staining seen has a cytoplasmic pattern in infected cells. It is also effective in paraffin embedded material. This antibody is anti morbillivirus specific and reacts with phocine distemper virus, racoon distemper virus and human measles virus. It does not cross react with CAV2, CCV, CPI or CPV.
Target Species Virus
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.