Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Canine Parvovirus Antibody (7341), Novus Biologicals™
SDP

Catalog No. NB10062433 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10062433 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10062433 Supplier Novus Biologicals Supplier No. NB10062433

Mouse Monoclonal Antibody

Canine Parvovirus Monoclonal specifically detects Canine Parvovirus in Virus samples. It is validated for ELISA.

Specifications

Antigen Canine Parvovirus
Applications ELISA
Classification Monoclonal
Clone 7341
Conjugate Unconjugated
Dilution ELISA 1:20-1:200
Gene Alias CPV, Parvo
Host Species Mouse
Immunogen Canine parvovirus
Purification Method Affinity Purified
Quantity 0.05 mg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This product detects Canine parvovirus (CPV), a negatively stranded DNA virus which infects dogs. In some instances it is also known as CPV type-2 to distinguish it from the unrelated parvovirus minute virus of canines (MVC). CPV is a relatively new virus, having been first isolated in 1978, which is thought to have derived from the feline panleukopenia virus (FPV). CPV is highly infectious and is spread from dog to dog by physical contact and contact with feces.
Target Species Virus
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.