Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Canine Parvovirus Antibody (8H7), Janelia Fluor™ 646, Novus Biologicals™
SDP

Catalog No. NB359964 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB359964 0.2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB359964 Supplier Novus Biologicals Supplier No. NB10073204JF646
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Canine Parvovirus Monoclonal specifically detects Canine Parvovirus in Virus samples. It is validated for Haemagglutination, Immunodiffusion, Lateral Flow Assay, Sandwich ELISA.

Specifications

Antigen Canine Parvovirus
Applications Hemagglutination Assay, Immunodiffusion, Lateral Flow, Sandwich ELISA
Classification Monoclonal
Clone 8H7
Conjugate Janelia Fluor 646
Dilution Immunodiffusion, Haemagglutination, Sandwich ELISA, Lateral Flow Assay
Formulation 50mM Sodium Borate
Gene Alias CPV, Parvo
Host Species Mouse
Immunogen Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with Canine parvovirus.
Purification Method Protein A purified
Quantity 0.2 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.