Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cannabinoid R1/CB1/CNR1 Antibody (2A1), Novus Biologicals™
SDP

Catalog No. NB297681 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB297681 100 μL
NB297680 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB297681 Supplier Novus Biologicals Supplier No. NBP326648100UL
Add to Cart
Add to Cart

Human Monoclonal Antibody

Cannabinoid R1/CB1/CNR1 Monoclonal antibody specifically detects Cannabinoid R1/CB1/CNR1 in Human samples. It is validated for ELISA

Specifications

Antigen Cannabinoid R1/CB1/CNR1
Applications ELISA
Classification Monoclonal
Clone 2A1
Conjugate Unconjugated
Dilution ELISA
Formulation 50% Glycerol, 0.01M PBS, PH 7.4
Gene Alias CANN6CB1R, cannabinoid receptor 1 (brain), CB1A, CB1K5, CB-RCB1cannabinoid receptor 1, central cannabinoid receptor, CNR
Host Species Human
Immunogen Recombinant Human Cannabinoid R1/CB1/CNR1 protein [UniProt P21554]
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Cancer, GPCR, Neuroscience, Neurotransmission, Vision
Primary or Secondary Primary
Gene ID (Entrez) 1268
Target Species Human
Content And Storage Store at -20 to -70C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG4
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.