Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cannabinoid R1/CB1/CNR1 Antibody (2A1), Novus Biologicals™

Human Monoclonal Antibody
$302.50 - $494.50
Specifications
Antigen | Cannabinoid R1/CB1/CNR1 |
---|---|
Clone | 2A1 |
Dilution | ELISA |
Applications | ELISA |
Classification | Monoclonal |
Description
Cannabinoid R1/CB1/CNR1 Monoclonal antibody specifically detects Cannabinoid R1/CB1/CNR1 in Human samples. It is validated for ELISASpecifications
Cannabinoid R1/CB1/CNR1 | |
ELISA | |
Monoclonal | |
Purified | |
RUO | |
Human | |
CANN6CB1R, cannabinoid receptor 1 (brain), CB1A, CB1K5, CB-RCB1cannabinoid receptor 1, central cannabinoid receptor, CNR | |
Recombinant Human Cannabinoid R1/CB1/CNR1 protein [UniProt P21554] | |
Primary | |
Store at -20 to -70C. Avoid freeze-thaw cycles. |
2A1 | |
ELISA | |
Unconjugated | |
Human | |
Cancer, GPCR, Neuroscience, Neurotransmission, Vision | |
50% Glycerol, 0.01M PBS, PH 7.4 | |
1268 | |
IgG4 | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title