Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cannabinoid R1/CB1/CNR1 Antibody (368302), Allophycocyanin/Cy7, Novus Biologicals™
SDP

Catalog No. NB312708 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB312708 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB312708 Supplier Novus Biologicals Supplier No. FAB3834APCCY7
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Cannabinoid R1/CB1/CNR1 Monoclonal specifically detects Cannabinoid R1/CB1/CNR1 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen Cannabinoid R1/CB1/CNR1
Applications Flow Cytometry
Classification Monoclonal
Clone 368302
Conjugate APC-Cyanine7
Dilution Flow Cytometry
Formulation PBS
Gene Alias CANN6CB1R, cannabinoid receptor 1 (brain), CB1A, CB1K5, CB-RCB1cannabinoid receptor 1, central cannabinoid receptor, CNR
Host Species Mouse
Immunogen HEK293 human embryonic kidney cell line transfected with human Cannabinoid R1/CB1/CNR1, Met1-Leu472, Accession # NP_057167
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, GPCR, Neuroscience, Neurotransmission, Vision
Primary or Secondary Primary
Gene ID (Entrez) 1268
Target Species Human
Content And Storage Store at 4C in the dark. Do not freeze.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.