Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CAP1 Antibody, Janelia Fluor™ 549, Novus Biologicals™
SDP

Catalog No. NB061640 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB061640 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB061640 Supplier Novus Biologicals Supplier No. NBP299569JF549
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CAP1 Polyclonal specifically detects CAP1 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.

Specifications

Antigen CAP1
Applications Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Conjugate Janelia Fluor 549
Dilution Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias adenylyl cyclase-associated protein 1, CAP, adenylate cyclase-associated protein 1 (yeast), CAP1-PEN, CAPCAP 1, SSKAP55
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the center region of human CAP1.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 10487
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.