Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CapG Antibody, HRP, Novus Biologicals™
SDP

Catalog No. NB065144 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB065144 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB065144 Supplier Novus Biologicals Supplier No. NBP300258H
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CapG Polyclonal specifically detects CapG in Human samples. It is validated for Western Blot, ELISA.

Specifications

Antigen CapG
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate HRP
Dilution Western Blot, ELISA
Formulation PBS
Gene Alias actin-regulatory protein CAP-G, AFCP, capping protein (actin filament), gelsolin-like, gelsolin-like capping protein, macrophage capping protein, macrophage-capping protein, MCPActin regulatory protein CAP-G
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant CapG (AAH14549.1; Met1-Lys348).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology, Cellular Markers
Primary or Secondary Primary
Gene ID (Entrez) 822
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.