Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Capicua Antibody, Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. N110990A70 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N110990A70 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N110990A70 Supplier Novus Biologicals Supplier No. NB11059905AF750
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Capicua Polyclonal specifically detects Capicua in Human, Mouse, Rat, Canine samples. It is validated for Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Capicua
Applications Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Conjugate Alexa Fluor 750
Dilution Immunocytochemistry/Immunofluorescence
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias capicua homolog (Drosophila), KIAA0306capicua (Drosophila) homolog, protein capicua homolog
Gene Symbols CIC
Host Species Rabbit
Immunogen The epitope recognized by this antibody maps to a region within residue 1150-1200 of human Capicua. [NP_055940.3]
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 23152
Target Species Human, Mouse, Rat, Canine
Content And Storage Store at 4°C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.