Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Capicua Antibody, Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB339154 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB339154 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB339154 Supplier Novus Biologicals Supplier No. NB11059905APC
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Capicua Polyclonal antibody specifically detects Capicua in Human, Mouse, Rat, Canine samples. It is validated for Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Western Blot (Negative)

Specifications

Antigen Capicua
Applications Immunocytochemistry, Immunoprecipitation
Classification Polyclonal
Conjugate APC
Dilution Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Western Blot (Negative)
Formulation PBS
Gene Alias capicua homolog (Drosophila), KIAA0306capicua (Drosophila) homolog, protein capicua homolog
Host Species Rabbit
Immunogen The epitope recognized by this antibody maps to a region within residue 1150-1200 of human Capicua. [NP_055940.3]
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 23152
Target Species Human, Mouse, Rat, Canine
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.