Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Capicua Antibody, Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB339148 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB339148 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB339148 Supplier Novus Biologicals Supplier No. NB11059906APC

Rabbit Polyclonal Antibody

Capicua Polyclonal antibody specifically detects Capicua in Human, Mouse, Rat, Canine samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, KnockDown

Specifications

Antigen Capicua
Applications Western Blot, Immunocytochemistry, Immunoprecipitation, KnockDown
Classification Polyclonal
Conjugate APC
Dilution Western Blot, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Knockout Validated
Formulation PBS
Gene Alias capicua homolog (Drosophila), KIAA0306capicua (Drosophila) homolog, protein capicua homolog
Host Species Rabbit
Immunogen The epitope recognized by this antibody maps to a C-terminal region within residue 1500-1608 of human Capicua. [NP_055940.3]
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 23152
Target Species Human, Mouse, Rat, Canine
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.