Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Capicua Antibody, DyLight 488, Novus Biologicals™
SDP

Catalog No. NB11059905G Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB11059905G 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB11059905G Supplier Novus Biologicals Supplier No. NB11059905G
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Capicua Polyclonal specifically detects Capicua in Human, Mouse, Rat, Canine samples. It is validated for Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Capicua
Applications Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Conjugate DyLight 488
Dilution Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Accession No. Q96RK0
Gene Alias capicua homolog (Drosophila), KIAA0306capicua (Drosophila) homolog, protein capicua homolog
Gene Symbols CIC
Host Species Rabbit
Immunogen The epitope recognized by this antibody maps to a region within residue 1150-1200 of human Capicua. [NP_055940.3]
Molecular Weight of Antigen 164 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 23152
Target Species Human, Mouse, Rat, Canine
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.