Learn More
Description
- Simplifies sample identification
- Press into place
- Frosted writing area on top provides space for additional identifying markings
- Made of polypropylene
Order Info
Available in 11 colors. Assorted packages contain 100 of each color.
Specifications
Specifications
| Pre-sterilized | No |
| Autoclavable | Autoclavable |
| Material | Polypropylene |
| Color | Gray |
| For Use With (Application) | Used to color code and write on frosted area for sample identification |
| No. per Pack | 500 Pk. |
| Sterility | Non-sterile |
| Tube Material | Polypropylene |
| For Use With (Equipment) | Micrewtubes Microcentrifuge Tubes and Other Products |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.