Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Simport™ Scientific Capinsert™ Cap Inserts

Catalog No. 22045423 Shop All Simport Scientific Products
Change view
Click to view available options
Color:
Assorted (Blue, Lilac, Red, Yellow, White)
Gray
Green
Orange
Pink
Red
Violet
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
22-045-423 Violet
22-045-361 Green
22-045-363 Orange
22-045-360 Gray
22-045-422 Red
22-045-421 Pink
22-045-424 Assorted (Blue, Lilac, Red, Yellow, White)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 22-045-423 Supplier Simport™ Scientific Supplier No. T345V
Add to Cart
Add to Cart

Capinsert™ Cap Inserts are used to color code a multitude of products such as tubes and vials. They are inserted on top of the closure and has a write-on frosted area for sample identification.

  • Simplifies sample identification
  • Press into place
  • Frosted writing area on top provides space for additional identifying markings
  • Made of polypropylene

Order Info

Available in 11 colors. Assorted packages contain 100 of each color.

Specifications

Pre-sterilized No
Autoclavable Autoclavable
Material Polypropylene
Color Violet
For Use With (Application) Used to color code and write on frosted area for sample identification
No. per Pack 500 Pk.
Sterility Non-sterile
Tube Material Polypropylene
For Use With (Equipment) Micrewtubes Microcentrifuge Tubes and Other Products
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.