Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NLRP3/NALP3 Antibody (Nalpy3-b) - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
$464.00
Specifications
Applications | Western Blot |
---|---|
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Regulatory Status | RUO |
Description
Zinc finger protein pseudogene Polyclonal specifically detects Zinc finger protein pseudogene in Human samples. It is validated for Western Blot.Specifications
Western Blot | |
Unconjugated | |
RUO | |
728743 | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
Polyclonal | |
Rabbit | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
IgG | |
Affinity Purified | |
22 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title