Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Capsaicin, USP, Spectrum™ Chemical
SDP

Catalog No. 18601420 Shop All Spectrum Chemical Manufacturing Corporation Products
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
1 kg
Packaging:
Amber Glass Bottle

Catalog No. 18-601-420

Supplier: Spectrum Chemical Manufacturing Corporation C18691GM

Only null left
Add to Cart
Add to Cart

C1869, 404-86-4, C18H27NO3

Spectrum™ Chemical Capsaicin, USP is used as an analgesic in nasal spays, topical ointments and in dermal patches to relieve pain. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 404-86-4
Molecular Formula C18H27NO3
Molecular Weight (g/mol) 305.42
InChI Key YKPUWZUDDOIDPM-SOFGYWHQSA-N
IUPAC Name (6E)-N-[(4-hydroxy-3-methoxyphenyl)methyl]-8-methylnon-6-enamide
SMILES COC1=CC(CNC(=O)CCCC\C=C\C(C)C)=CC=C1O

Specifications

CAS Max % 100%
Ignition Residue 1%
Loss on Drying 1%
Melting Range 57-66°C
Packaging Amber Glass Bottle
Quantity 1 g
UN Number UN2811
Percent Purity 90 to 110%
Grade USP
Physical Form Crystalline Powder or Lumps
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.