Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CAPZA1 Mouse anti-Human, Mouse, Rat, Clone: 10H1F9, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
Antigen | CAPZA1 |
---|---|
Clone | 10H1F9 |
Concentration | 0.85 mg/mL |
Applications | Immunocytochemistry, Immunofluorescence, Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-6503
![]() ![]() |
Proteintech Group Inc
660661IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
CAPZA1 is a member of the F-actin capping protein alpha subunit family. This gene encodes the alpha subunit of the barbed-end actin binding protein. The protein regulates growth of the actin filament by capping the barbed end of growing actin filaments.Specifications
CAPZA1 | |
0.85 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
CAPPA1, CAPZ, CapZ alpha 1, CAPZA1, CAZ1 | |
CAPZA1 | |
IgG1 | |
Protein G | |
Antibody |
10H1F9 | |
Immunocytochemistry, Immunofluorescence, Western Blot | |
Unconjugated | |
Mouse | |
Human, Mouse, Rat | |
B2GUZ5, P47753, P52907 | |
12340, 691149, 829 | |
CAPZA1 Fusion Protein Ag18841 | |
Primary | |
-20°C | |
CAPZA1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title