Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CAPZB Antibody, Novus Biologicals™
SDP

Catalog No. NB100814 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB100814 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB100814 Supplier Novus Biologicals Supplier No. NB100814
Add to Cart
Add to Cart

Goat Polyclonal Antibody

CAPZB Polyclonal specifically detects CAPZB in Human, Mouse, Pig, Rat samples. It is validated for Peptide ELISA, Western Blot.

Specifications

Antigen CAPZB
Applications Peptide ELISA, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.1 - 0.3 ug/ml, Peptide ELISA Detection limit 1:32000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. NP_004921.1
Gene Alias Cap Z, CAPB, CAPPB, capping protein (actin filament) muscle Z-line, beta, CAPZ, CapZ beta, F-actin capping protein beta subunit, F-actin-capping protein subunit beta, MGC104401, MGC129749, MGC129750
Gene Symbols CAPZB
Host Species Goat
Immunogen Peptide with sequence NDLVEALKRKQQC corresponding to C-Terminus according to NP_004921.1.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 832
Test Specificity This antibody is expected to be specific for the beta2 isoform of capping protein, found in non-muscle cells and should not recognize the beta1 form.
Target Species Human, Mouse, Rat, Pig, Rat
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.