Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Capzimin dimer
GREENER_CHOICE

Catalog No. p-7250118
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Selective proteasome inhibitor

Capzimin dimer is a specific inhibitor of proteasome isopeptidase Rpn11 (IC50 = 2.1 μM and 3.8 μM, in 293T and A549 cell lines, respectively). It exhibits 80-fold selectivity for Rpn11 over Csn5, ten-fold over AMSH and six-fold over BRCC36 (IC50 = 30 μM, 4.5 μM, and 2.3 μM, respectively). Capzimin dimer blocks proliferation of cancer cells.

Specifications

Chemical Name or Material 8,8 '-Dithiobis[N-[2-(2-thiazolyl)ethyl]-3-quinolinecarboxamide]
CAS 2084868-04-0
Quantity 10 mg
Target Proteasome Inhibitors
Molecular Formula C30H24N6O2S4
Purity 0.98
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.