Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

carabin Antibody, Janelia Fluor™ 549, Novus Biologicals™
SDP

Catalog No. NB196546 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB196546 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB196546 Supplier Novus Biologicals Supplier No. NBP176806JF549
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

carabin Polyclonal antibody specifically detects carabin in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)

Specifications

Antigen carabin
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Janelia Fluor 549
Formulation 50mM Sodium Borate
Gene Alias carabin, FLJ00332, MGC46488, TBC1 domain family member 10C, TBC1 domain family, member 10C
Host Species Rabbit
Immunogen Antibody was raised against a 16 amino acid synthetic peptide from near the carboxy terminus of human Carabin. The immunogen is located within the last 50 amino acids of Carabin. Amino Acid Squence: PEEPRPPRRKPQTRGKT
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 374403
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.