Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Carbohydrate Sulfotransferase 2/CHST2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP15994420UL
Description
Carbohydrate Sulfotransferase 2/CHST2 Polyclonal specifically detects Carbohydrate Sulfotransferase 2/CHST2 in Human samples. It is validated for Western Blot.Specifications
Carbohydrate Sulfotransferase 2/CHST2 | |
Polyclonal | |
Western Blot 1:100-1:2000 | |
Q9Y4C5 | |
CHST2 | |
Synthetic peptides corresponding to CHST2(carbohydrate (N-acetylglucosamine-6-O) sulfotransferase 2) The peptide sequence was selected from the middle region of CHST2. Peptide sequence NAWRTALTFQQIKQVEEFCYQPMAVLGYERVNSPEEVKDLSKTLLRKPRL. | |
20 μL | |
Primary | |
Human | |
IgG |
Western Blot | |
Unconjugated | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
C6ST, carbohydrate (chondroitin 6/keratan) sulfotransferase 2, carbohydrate (N-acetylglucosamine-6-O) sulfotransferase 2, carbohydrate sulfotransferase 2, EC 2.8.2.-, Galactose/N-acetylglucosamine/N-acetylglucosamine 6-O-sulfotransferase 2, glcNAc6ST-1, GN6ST, Gn6ST-1, GST2, GST-2, N-acetylglucosamine 6-O-sulfotransferase 1 | |
Rabbit | |
Affinity Purified | |
RUO | |
9435 | |
Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction