Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carbohydrate Sulfotransferase 6/CHST6 Antibody (OTI1D1), DyLight 550, Novus Biologicals™
SDP

Catalog No. NBP271195R Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP271195R 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP271195R Supplier Novus Biologicals Supplier No. NBP271195R
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Carbohydrate Sulfotransferase 6/CHST6 Monoclonal specifically detects Carbohydrate Sulfotransferase 6/CHST6 in Human, Mouse, Canine, Monkey samples. It is validated for Western Blot.

Specifications

Antigen Carbohydrate Sulfotransferase 6/CHST6
Applications Western Blot
Classification Monoclonal
Clone OTI1D1
Conjugate DyLight 550
Dilution Western Blot
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias carbohydrate (N-acetylglucosamine 6-O) sulfotransferase 6, carbohydrate sulfotransferase 6, C-GlcNAc6ST, corneal N-acetylglucosamine 6-sulfotransferase, Corneal N-acetylglucosamine-6-O-sulfotransferase, EC 2.8.2, EC 2.8.2.-, EC 2.8.2.17, EC 2.8.2.21, Galactose/N-acetylglucosamine/N-acetylglucosamine 6-O-sulfotransferase 4-beta, glcNAc6ST-5, Gn6st-5, GST4-beta, hCGn6ST, MCDC1, N-acetylglucosamine 6-O-sulfotransferase 5
Gene Symbols CHST6
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 202-395 of human Carbohydrate Sulfotransferase 6/CHST6 (NP_067628) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 4166.0
Target Species Human, Mouse, Canine, Monkey
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.