Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ Carbon / Technora Full Face Hood

Catalog No. p-7180083 Shop All Protective Industrial Products Inc. Products
Click to view available options
Design Type:
Straight Cut Design
Tri-Cut Design
This item is not returnable. View return policy
This item is not returnable. View return policy

PIP™ Carbon / Technora Full Face Hood features lightweight carbon fiber for excellent fire resistance

  • Carbon fiber is lightweight and provides excellent fire resistance
  • Technora™ adds strength, durability, and helps hood retain its shape when launder
  • Full face coverage
  • TPP Rating 28
  • Meets NFPA 1971 Requirements
  • One Size Fits Most

Specifications

Product Type Fire Protectant Hood
Size Universal
Material Nomex
Color Black
Disposable Reusable
Design Type Straight Cut Design
Quantity 1 Ea.
Includes Hood Only
For Use With (Application) Heat/Fire Applications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.