Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carbonic Anhydrase II/CA2 Antibody (003), DyLight 755, Novus Biologicals™
SDP

Catalog No. NB040561 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB040561 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB040561 Supplier Novus Biologicals Supplier No. NBP290677IR
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Carbonic Anhydrase II/CA2 Monoclonal specifically detects Carbonic Anhydrase II/CA2 in Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Carbonic Anhydrase II/CA2
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 3
Conjugate DyLight 755
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias CAII, CA-IIEC 4.2.1.1, Car2, Carbonate dehydratase II, carbonic anhydrase 2, carbonic anhydrase B, Carbonic anhydrase C, carbonic anhydrase IICAC, carbonic dehydratase
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Mouse Carbonic Anhydrase II/CA2 (AAH55291.1; Met1-Lys260).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Hypoxia, Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 760
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.