Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carbonic Anhydrase III/CA3 Antibody (002), DyLight 550, Novus Biologicals™
SDP

Catalog No. NB108218 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB108218 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB108218 Supplier Novus Biologicals Supplier No. NBP289537R
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Carbonic Anhydrase III/CA3 Monoclonal specifically detects Carbonic Anhydrase III/CA3 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Carbonic Anhydrase III/CA3
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 2
Conjugate DyLight 550
Formulation 50 mM Sodium Borate
Gene Alias CAIII, CA-III, Car3, Carbonate dehydratase III, carbonic anhydrase 3, Carbonic anhydrase III, carbonic anhydrase III, muscle specific, EC 4.2.1.1
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human Carbonic Anhydrase III/CA3 (NP_005172.1; Met1-Lys260).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 761
Test Specificity No cross-reactivity in ELISA with: Human CA2 Human CA4 Human CA5a Human CA5b Human CA7 Human CA8 Human CA9 Human CA10 Human CA13 Human CA14
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.