Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carbonic Anhydrase IV/CA4 Antibody (039), FITC, Novus Biologicals™
SDP

Catalog No. NB108077 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB108077 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB108077 Supplier Novus Biologicals Supplier No. NBP289518F
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Carbonic Anhydrase IV/CA4 Monoclonal specifically detects Carbonic Anhydrase IV/CA4 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Carbonic Anhydrase IV/CA4
Applications ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 39
Conjugate FITC
Formulation PBS
Gene Alias CAIV, CA-IV, Car4, Carbonate dehydratase IV, carbonic anhydrase 4, carbonic anhydrase IVRP17, carbonic dehydratase IV, EC 4.2.1.1, retinitis pigmentosa 17 (autosomal dominant)
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human Carbonic Anhydrase IV/CA4 (NP_000708.1; Met1-Lys283).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Lipid and Metabolism, Vision
Primary or Secondary Primary
Gene ID (Entrez) 762
Test Specificity No cross-reactivity in ELISA with: Human CA2 Human CA3 Human CA5A Human CA5B Human CA8 Human CA9 Human CA10 Human CA12 Human CA13 Human CA14
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.