Learn More
Carbonic Anhydrase IX/CA9 Antibody (2D3), Allophycocyanin, Novus Biologicals™
Shop All R&D Systems Products

Description
Specifications
Specifications
| Antigen | Carbonic Anhydrase IX/CA9 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 2D3 |
| Conjugate | APC |
| Dilution | Flow Cytometry, Flow (Intracellular) |
| Gene Accession No. | Q16790 |
| Gene Alias | CA-IX, CAIXcarbonic anhydrase 9, Carbonate dehydratase IX, carbonic anhydrase IXpMW1, carbonic dehydratase, EC 4.2.1.1, G250, Membrane antigen MN, MNRCC-associated antigen G250, P54/58N, RCC-associated protein G250, Renal cell carcinoma-associated antigen G250 |
| Gene Symbols | CA9 |
| Host Species | Mouse |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.