Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carbonic Anhydrase IX/CA9 Antibody (303123), PE/Cy7, Novus Biologicals™
SDP

Catalog No. NB313697 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB313697 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB313697 Supplier Novus Biologicals Supplier No. FAB2188PECY7
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Carbonic Anhydrase IX/CA9 Monoclonal specifically detects Carbonic Anhydrase IX/CA9 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen Carbonic Anhydrase IX/CA9
Applications Flow Cytometry
Classification Monoclonal
Clone 303123
Conjugate PE-Cyanine7
Dilution Flow Cytometry
Formulation PBS
Gene Alias CAIX, CA-IX, Carbonate dehydratase IX, carbonic anhydrase 9, carbonic anhydrase IX, carbonic dehydratase, EC 4.2.1.1, G250, Membrane antigen MN, MN, P54/58N, PMW1, RCC-associated antigen G250, RCC-associated protein G250, Renal cell carcinoma-associated antigen G250
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human Carbonic Anhydrase IX, Pro59-Asp414, Accession # Q16790
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, HIF Target Genes, Hypoxia, Lipid and Metabolism, Metastasis, Renal Cell Carcinoma Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 768
Target Species Human
Content And Storage Store at 4C in the dark. Do not freeze.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.