Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carbonic Anhydrase IX/CA9 Antibody (4F12), Novus Biologicals™
SDP

Catalog No. NB298040 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB298040 50 μL
NB298041 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB298040 Supplier Novus Biologicals Supplier No. NBP32646850UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Carbonic Anhydrase IX/CA9 Monoclonal antibody specifically detects Carbonic Anhydrase IX/CA9 in Human samples. It is validated for ELISA, Immunohistochemistry

Specifications

Antigen Carbonic Anhydrase IX/CA9
Applications ELISA, Immunohistochemistry
Classification Monoclonal
Clone 4F12
Conjugate Unconjugated
Dilution ELISA, Immunohistochemistry 1:50-1:200
Formulation PBS, pH 7.4, 150mM NaCl and 50% glycerol
Gene Alias CAIX, CA-IX, Carbonate dehydratase IX, carbonic anhydrase 9, carbonic anhydrase IX, carbonic dehydratase, EC 4.2.1.1, G250, Membrane antigen MN, MN, P54/58N, PMW1, RCC-associated antigen G250, RCC-associated protein G250, Renal cell carcinoma-associated antigen G250
Host Species Rabbit
Immunogen A synthesized peptide derived from Human Carbonic Anhydrase IX/CA9 [UniProt Q16790]
Purification Method Affinity purified
Quantity 50 μL
Regulatory Status RUO
Research Discipline Cancer, HIF Target Genes, Hypoxia, Lipid and Metabolism, Metastasis, Renal Cell Carcinoma Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 768
Target Species Human
Content And Storage Store at -20 to -70C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.