Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carbonic Anhydrase IX/CA9 Antibody, Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NB209642 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB209642 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB209642 Supplier Novus Biologicals Supplier No. NB100417AF532
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Carbonic Anhydrase IX/CA9 Polyclonal specifically detects Carbonic Anhydrase IX/CA9 in Human, Mouse, Plant, Rat samples. It is validated for ChIP Assay, Dual RNAScope ISH-IHC, ELISA, Flow Cytometry, Gel Super Shift Assays, Immunoblot, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Microarray, Proximity Ligation Assay, Western Blot.

Specifications

Antigen Carbonic Anhydrase IX/CA9
Applications ChIP Assay, Dual RNAScope ISH-IHC, ELISA, Flow Cytometry, Gel Supershift, Immunoblot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Microarray, Proximity Ligation Assay, Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 532
Formulation 50mM Sodium Borate
Gene Alias CAIX, CA-IX, Carbonate dehydratase IX, carbonic anhydrase 9, carbonic anhydrase IX, carbonic dehydratase, EC 4.2.1.1, G250, Membrane antigen MN, MN, P54/58N, PMW1, RCC-associated antigen G250, RCC-associated protein G250, Renal cell carcinoma-associated antigen G250
Host Species Rabbit
Immunogen This Carbonic Anhydrase IX/CA9 Antibody was made from a synthetic peptide from the C-terminal sequence of human Carbonic Anhydrase IX (within residues 400-459) [UniProt# Q16790].
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, HIF Target Genes, Hypoxia, Lipid and Metabolism, Metastasis, Renal Cell Carcinoma Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 768
Target Species Human, Mouse, Mouse, Plant, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.