Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carbonic Anhydrase IX/CA9 Antibody (CA9/781), Novus Biologicals™
SDP

Catalog No. NBP24523702 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.02 mg
0.1 mg
0.2 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP24523702 0.2 mg
NBP24523700 0.02 mg
NBP24523701 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. NBP24523702 Supplier Novus Biologicals Supplier No. NBP2452370.2MG

Mouse Monoclonal Antibody

Ensure accurate, reproducible results in Western Blot, Flow Cytometry, Immunohistochemistry (Paraffin), Immunofluorescence

Carbonic Anhydrase IX/CA9 Monoclonal antibody specifically detects Carbonic Anhydrase IX/CA9 in Human, Equine samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry-Paraffin, SDS-Page, Immunofluorescence.

Specifications

Antigen Carbonic Anhydrase IX/CA9
Applications Western Blot, Flow Cytometry, Immunohistochemistry (Paraffin), SDS-Page, Immunofluorescence
Classification Monoclonal
Clone CA9/781
Concentration 0.2mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.5 - 1.0 ug/ml, Flow Cytometry 0.5 - 1 ug/million cells in 0.1 ml, Immunohistochemistry-Paraffin 0.5 - 1.0 ug/ml, SDS-Page, Immunofluorescence 1 - 2 ug/ml
Formulation 10mM PBS and 0.05% BSA with 0.05% Sodium Azide
Gene Accession No. Q16790
Gene Alias CA-IX, CAIXcarbonic anhydrase 9, Carbonate dehydratase IX, carbonic anhydrase IXpMW1, carbonic dehydratase, EC 4.2.1.1, G250, Membrane antigen MN, MNRCC-associated antigen G250, P54/58N, RCC-associated protein G250, Renal cell carcinoma-associated antigen G250
Gene Symbols CA9
Host Species Mouse
Immunogen Recombinant human CA9 protein
Molecular Weight of Antigen 55 kDa
Purification Method Protein A or G purified
Quantity 0.2 mg
Regulatory Status RUO
Research Discipline Cancer, Cellular Markers, HIF Target Genes, Hypoxia, Lipid and Metabolism, Signal Transduction, Vision
Primary or Secondary Primary
Gene ID (Entrez) 768
Test Specificity Recognizes a glycoprotein of ∼200kDa, identified as carbonic anhydrase IX (CAIX/gp200). Carbonic Anhydrases (CAs) are members of a large family of zinc metallo-enzymes that catalyze the reversible hydration of carbon dioxide. CAs are involved in a variety of biological processes, including respiration, calcification, acid-base balance, bone resorption and the formation of aqueous humor, cerebrospinal fluid, saliva and gastric juice. They show extensive diversity in distribution and in their subcellular localization. CA IX is specifically expressed in clear-cell renal carcinomas.
Target Species Human, Equine
Content And Storage Store at 4C.
Form Purified
Isotype IgG2b κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.