Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carbonic Anhydrase IX/CA9 Antibody, DyLight 550, Novus Biologicals™
SDP

Catalog No. NB100417R Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB100417R 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB100417R Supplier Novus Biologicals Supplier No. NB100417R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Carbonic Anhydrase IX/CA9 Polyclonal specifically detects Carbonic Anhydrase IX/CA9 in Human, Mouse, Plant, Rat samples. It is validated for ChIP Assay, Dual RNAScope ISH-IHC, ELISA, Flow Cytometry, Gel Super Shift Assays, Immunoblot, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Microarray, Proximity Ligation Assay, Western Blot.

Specifications

Antigen Carbonic Anhydrase IX/CA9
Applications ChIP Assay, Dual RNAScope ISH-IHC, ELISA, Flow Cytometry, Gel Supershift, Immunoblot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunoprecipitation, Microarray, Proximity Ligation Assay, Western Blot
Classification Polyclonal
Conjugate DyLight 550
Dilution Western Blot, Flow Cytometry 1:1000, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunoblotting
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Accession No. Q16790
Gene Alias CA-IX, CAIXcarbonic anhydrase 9, Carbonate dehydratase IX, carbonic anhydrase IXpMW1, carbonic dehydratase, EC 4.2.1.1, G250, Membrane antigen MN, MNRCC-associated antigen G250, P54/58N, RCC-associated protein G250, Renal cell carcinoma-associated antigen G250
Gene Symbols CA9
Host Species Rabbit
Immunogen A synthetic peptide made to a C-terminal sequence of human Carbonic Anhydrase IX (within residues 400-459) [UniProt Q16790]
Molecular Weight of Antigen 55 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, HIF Target Genes, Hypoxia, Lipid and Metabolism, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 768
Test Specificity Human, dog, rat and mouse (PMID 22842475).
Target Species Human, Mouse, Rat, Plant, Rat
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.