Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carbonic Anhydrase VA/CA5A Antibody, PerCP, Novus Biologicals™
SDP

Catalog No. NB325997 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB325997 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB325997 Supplier Novus Biologicals Supplier No. NBP299341PCP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Carbonic Anhydrase VA/CA5A Polyclonal antibody specifically detects Carbonic Anhydrase VA/CA5A in Human samples. It is validated for Western Blot, ELISA

Specifications

Antigen Carbonic Anhydrase VA/CA5A
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate PerCP
Dilution Western Blot, ELISA
Formulation PBS
Gene Alias CA5carbonic anhydrase 5A, mitochondrial, Carbonate dehydratase VA, carbonic anhydrase V, mitochondrial, Carbonic anhydrase VA, carbonic anhydrase VA, mitochondrial, carbonic dehydratase, CAV, CAVA, CA-VA, EC 4.2.1.1
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Human Carbonic Anhydrase VA/CA5A (Accession#: NP_001730.1; Ala 40-Ser 305)
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 763
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.