Learn More
Description
Specifications
Specifications
| Antigen | Carbonic Anhydrase VB/CA5B |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | 30 |
| Conjugate | FITC |
| Dilution | ELISA |
| Formulation | PBS |
| Gene Alias | Carbonate dehydratase VB, carbonic anhydrase 5B, mitochondrial, Carbonic anhydrase VB, carbonic anhydrase VB, mitochondrial, carbonic dehydratase, CA-VB, EC 4.2.1.1, MGC39962 |
| Host Species | Rabbit |
| Immunogen | This antibody was obtained from a rabbit immunized with purified, recombinant Human Carbonic Anhydrase VB/CA5B (Q9Y2D0; Cys34-Pro 317). |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
