Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carbonic Anhydrase VB/CA5B Rabbit anti-Human, Clone: 8, Novus Biologicals™
SDP

Catalog No. NB021643 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB021643 100 μg
NB021644 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB021643 Supplier Novus Biologicals Supplier No. NBP290090100UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Carbonic Anhydrase VB/CA5B Monoclonal specifically detects Carbonic Anhydrase VB/CA5B in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Carbonic Anhydrase VB/CA5B
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 8
Conjugate Unconjugated
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin 1:50-1:200
Formulation 0.2 um filtered solution in PBS
Gene Alias Carbonate dehydratase VB, carbonic anhydrase 5B, mitochondrial, Carbonic anhydrase VB, carbonic anhydrase VB, mitochondrial, carbonic dehydratase, CA-VB, EC 4.2.1.1, MGC39962
Host Species Rabbit
Immunogen Recombinant Human Carbonic Anhydrase VB/CA5B protein
Purification Method Protein A purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 11238
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.