Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carbonic Anhydrase VI Antibody (401809), mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB348623 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB348623 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB348623 Supplier Novus Biologicals Supplier No. FAB29391MFV450
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Carbonic Anhydrase VI Monoclonal antibody specifically detects Carbonic Anhydrase VI in Human samples. It is validated for Flow Cytometry

Specifications

Antigen Carbonic Anhydrase VI
Applications Flow Cytometry
Classification Monoclonal
Clone 401809
Conjugate mFluor Violet 450 SE
Dilution Intracellular Staining by Flow Cytometry
Formulation 50mM Sodium Borate
Gene Alias Carbonate dehydratase VI, carbonic anhydrase 6, carbonic anhydrase VIGUSTIN, CA-VI, EC 4.2.1.1, MGC21256, Salivary carbonic anhydrase, Secreted carbonic anhydrase
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human Carbonic Anhydrase VI, Gln18-Asn308, Accession # EAW71606
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 765
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.