Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carbonic Anhydrase VIII/CA8 Antibody (001), HRP, Novus Biologicals™
SDP

Catalog No. NB076772 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB076772 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB076772 Supplier Novus Biologicals Supplier No. NBP290709H
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Carbonic Anhydrase VIII/CA8 Monoclonal specifically detects Carbonic Anhydrase VIII/CA8 in Mouse samples. It is validated for ELISA, Sandwich ELISA Detection.

Specifications

Antigen Carbonic Anhydrase VIII/CA8
Applications ELISA
Classification Monoclonal
Clone 1
Conjugate HRP
Formulation PBS
Gene Alias CALScarbonate dehydratase, CAMRQ3, carbonic anhydrase VIIIMGC99509, carbonic anhydrase-like sequence, carbonic anhydrase-related protein, CARPCA-related protein, CA-VIII, MGC120502
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Mouse Carbonic Anhydrase VIII/CA8 (P28651; Met1-Gln291).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 767
Test Specificity Has cross-reactivity in ELISA with: Human CA8
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.