Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carbonic Anhydrase VIII/CA8 Antibody (008), Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. NB108054 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB108054 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB108054 Supplier Novus Biologicals Supplier No. NBP289517AF700
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Carbonic Anhydrase VIII/CA8 Monoclonal specifically detects Carbonic Anhydrase VIII/CA8 in Human samples. It is validated for Western Blot.

Specifications

Antigen Carbonic Anhydrase VIII/CA8
Applications Western Blot
Classification Monoclonal
Clone 8
Conjugate Alexa Fluor 700
Formulation 50 mM Sodium Borate
Gene Alias CALScarbonate dehydratase, CAMRQ3, carbonic anhydrase VIIIMGC99509, carbonic anhydrase-like sequence, carbonic anhydrase-related protein, CARPCA-related protein, CA-VIII, MGC120502
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human Carbonic Anhydrase VIII/CA8 (NP_004047.3; Met1-Gln290).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 767
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.