Learn More
Carbonic Anhydrase VIII/CA8 Rabbit anti-Mouse, Clone: 1, Novus Biologicals™
Shop All R&D Systems Products

Description
Specifications
Specifications
| Antigen | Carbonic Anhydrase VIII/CA8 |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | 1 |
| Conjugate | Unconjugated |
| Dilution | ELISA 1:5000-1:10000, Sandwich ELISA Detection 1:1000-1:10000 |
| Formulation | 0.2 um filtered solution in PBS |
| Gene Alias | CALScarbonate dehydratase, CAMRQ3, carbonic anhydrase VIIIMGC99509, carbonic anhydrase-like sequence, carbonic anhydrase-related protein, CARPCA-related protein, CA-VIII, MGC120502 |
| Host Species | Rabbit |
| Immunogen | Recombinant Mouse Carbonic Anhydrase VIII/CA8 protein |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.