Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carbonic Anhydrase X/CA10 Antibody (018), Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NB076781 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB076781 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB076781 Supplier Novus Biologicals Supplier No. NBP290712AF488
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Carbonic Anhydrase X/CA10 Monoclonal specifically detects Carbonic Anhydrase X/CA10 in Mouse samples. It is validated for ELISA.

Specifications

Antigen Carbonic Anhydrase X/CA10
Applications ELISA
Classification Monoclonal
Clone 18
Conjugate Alexa Fluor 488
Formulation 50 mM Sodium Borate
Gene Alias carbonic anhydrase X, carbonic anhydrase-related protein 10, Carbonic anhydrase-related protein X, CARP X, CA-RPX, CARPXCA-RP X, Cerebral protein 15, cerebral protein-15, HUCEP-15
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Mouse Carbonic Anhydrase X/CA10 (P61215; Met1-Asn300).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56934
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.