Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carbonic Anhydrase X/CA10 Antibody, Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB321835 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB321835 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB321835 Supplier Novus Biologicals Supplier No. NBP298120APC
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Carbonic Anhydrase X/CA10 Polyclonal antibody specifically detects Carbonic Anhydrase X/CA10 in Mouse samples. It is validated for Western Blot, ELISA

Specifications

Antigen Carbonic Anhydrase X/CA10
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate APC
Dilution Western Blot, ELISA
Formulation PBS
Gene Alias carbonic anhydrase X, carbonic anhydrase-related protein 10, Carbonic anhydrase-related protein X, CARP X, CA-RPX, CARPXCA-RP X, Cerebral protein 15, cerebral protein-15, HUCEP-15
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Mouse Carbonic Anhydrase X/CA10 (Accession#: P61215; Met1-Asn300)
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56934
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.