Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carbonic Anhydrase XII/CA12 Antibody (08), Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NB292943 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB292943 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB292943 Supplier Novus Biologicals Supplier No. NBP326096AF594

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Mouse Monoclonal Antibody

Carbonic Anhydrase XII/CA12 Monoclonal antibody specifically detects Carbonic Anhydrase XII/CA12 in Human samples. It is validated for ELISA

Specifications

Antigen Carbonic Anhydrase XII/CA12
Applications ELISA
Classification Monoclonal
Clone 08
Conjugate Alexa Fluor 594
Dilution ELISA
Formulation 50mM Sodium Borate
Gene Alias Carbonate dehydratase XII, carbonic anhydrase 12, carbonic anhydrase XIICAXII, carbonic dehydratase, CA-XII, EC 4.2.1.1, FLJ20151, HsT18816, Tumor antigen HOM-RCC-3.1.3
Host Species Mouse
Immunogen This product is a recombinant monoclonal antibody expressed from HEK293 cells.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 771
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.