Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carbonic Anhydrase XII/CA12 Antibody (OTI2C6), Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NP270329A32 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP270329A32 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP270329A32 Supplier Novus Biologicals Supplier No. NBP270329AF532
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Carbonic Anhydrase XII/CA12 Monoclonal specifically detects Carbonic Anhydrase XII/CA12 in Human samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen Carbonic Anhydrase XII/CA12
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI2C6
Conjugate Alexa Fluor 532
Dilution Western Blot, Immunohistochemistry
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias Carbonate dehydratase XII, carbonic anhydrase 12, carbonic anhydrase XIICAXII, carbonic dehydratase, CA-XII, EC 4.2.1.1, FLJ20151, HsT18816, Tumor antigen HOM-RCC-3.1.3
Gene Symbols CA12
Host Species Mouse
Immunogen Full length recombinant protein of human Carbonic Anhydrase XII/CA12 (NP_001209) produced in HEK293T cells.
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 771
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.