Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Supelco™ Carbotrap™ 202 Thermal Desorption Tubes

Catalog No. 111006682 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Description:
Pre-conditioned
Pre-conditioned/Sealed with Brass Endcaps
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Description
11-100-6682 Pre-conditioned/Sealed with Brass Endcaps
11-100-6696 Pre-conditioned
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-100-6682 Supplier MilliporeSigma™ Supelco™ Supplier No. 29745U
Add to Cart
Add to Cart

Suitable for use with Air Monitoring

TRUSTED_SUSTAINABILITY

Specifications

Matrix Carbopack™ C Graphitized Carbon Black (GCB), Carbopack™ Graphitized Carbon Black (GCB), Carbotrap™ 202
Description Pre-conditioned/Sealed with Brass Endcaps
For Use With (Equipment) PerkinElmer, Markes, DANI, OI Analytical, and Shimadzu Instruments
Material Stainless Steel
Diameter (English) Outer 1/4 in.
For Use With (Application) Suitable for Air Monitoring (Compounds: C5-C20)
Product Line Carbotrap
Packaging Quantity 10/Pk.
Overall Length (English) 3.5 in.
Type Thermal Desorption Tube

Carbotrap is a registered trademark of Sigma-Aldrich Co. LLC

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.