Learn More
MilliporeSigma™ Supelco™ Carbotrap™ 300 Thermal Desorption Tubes
Shop All MilliporeSigma Supelco ProductsSpecifications
Specifications
| Methods | EPA TO-1,TO-2,TO-3,TO-17 |
| Description | Pre-conditioned |
| For Use With (Application) | Air Monitoring |
| Packaging Quantity | 10/Pk. |
| Product Line | Carbotrap |
| Matrix | Carbosieve™ S-III Carbon Molecular Sieve (CMS), Carbotrap™ 300, Carbotrap™ B Graphitized Carbon Black (GCB), Carbotrap™ C Graphitized Carbon Black (GCB) |
| Overall Length (English) | 3.5 in. |
| Type | Thermal Desorption Tube |
| Material | Stainless Steel |
| For Use With (Equipment) | PerkinElmer, Markes, DANI, OI Analytical, and Shimadzu Instruments |
| Show More |
Carbotrap is a registered trademark of Sigma-Aldrich Co. LLC
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.