Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carboxyl Ester Lipase/CEL Antibody (063), Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NB040425 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB040425 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB040425 Supplier Novus Biologicals Supplier No. NBP290647AF532
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Carboxyl Ester Lipase/CEL Monoclonal specifically detects Carboxyl Ester Lipase/CEL in Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Carboxyl Ester Lipase/CEL
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 63
Conjugate Alexa Fluor 532
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias BAL, bile salt-activated lipase, Bile salt-stimulated lipase, BSDL, BSSLbile salt-dependent lipase, oncofetal isoform, bucelipase, carboxyl ester hydrolase, Carboxyl ester lipase, carboxyl ester lipase (bile salt-stimulated lipase), CEase, CELL, Cholesterol esterase, EC 3.1.1, EC 3.1.1.13, EC 3.1.1.3, FAP, FAPP, fetoacinar pancreatic protein, LIPA, lysophospholipase, pancreatic, MODY8bile-salt-activated lipase, Pancreatic lysophospholipase, Sterol esterase
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Mouse Carboxyl Ester Lipase/CEL (NP_034015.1; Met1-Leu534).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1056
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.