Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carboxylesterase 1/CES1 Antibody, Novus Biologicals™
SDP

Catalog No. NBP244236 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP244236 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP244236 Supplier Novus Biologicals Supplier No. NBP244236
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifically detects Carboxylesterase 3/CES3/Esterase 31 in Porcine samples, and it is validated for ELISA, Western Blotting

Carboxylesterase 1/CES1 Polyclonal specifically detects Carboxylesterase 1/CES1 in Porcine samples. It is validated for Western Blot, ELISA.

Specifications

Antigen Carboxylesterase 1/CES1
Applications Western Blot, ELISA
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1:1000-1:5000, ELISA 1:10000-1:50000
Formulation 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide
Gene Accession No. Q29550
Gene Alias ACAT, acyl coenzyme A:cholesterol acyltransferase, Acyl-coenzyme A:cholesterol acyltransferase, Brain carboxylesterase hBr1, carboxylesterase 1, carboxylesterase 1 (monocyte/macrophage serine esterase 1), carboxylesterase 2 (liver), CEH, Ces-1, CES1A2, CES2EC 3.1.1.1, cholesteryl ester hydrolase, Cocaine carboxylesterase, EC 3.1.1, egasyn, HMSE1, HMSECES1A1, human monocyte/macrophage serine esterase 1, liver carboxylesterase 1, MGC117365, Monocyte/macrophage serine esterase, PCE-1, REH, Retinyl ester hydrolase, Serine esterase 1, Ses-1, SES1TGH, Triacylglycerol hydrolase
Gene Symbols CES1
Host Species Rabbit
Immunogen Carboxylesterase 3/CES3/Carboxylesterase 3/CES3/Esterase 31 31 [Porcine Liver] (Uniprot: Q29550)
Molecular Weight of Antigen 62.02 kDa
Purification Method Multi-step
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1066
Reconstitution Reconstitute with 100 ul deionized water (or equivalent)
Target Species Pig
Content And Storage Store lyophilized antibody at 4°C. Aliquot reconstituted liquid and store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.