Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carboxymethyl Lysine Antibody (CML26), Novus Biologicals™
SDP

Catalog No. NB119855 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB119855 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB119855 Supplier Novus Biologicals Supplier No. NBP311230
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Carboxymethyl Lysine Monoclonal specifically detects Carboxymethyl Lysine in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunoassay.

Specifications

Antigen Carboxymethyl Lysine
Applications Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoassay
Classification Monoclonal
Clone CML26
Concentration 0.1 mg/ml
Conjugate Unconjugated
Dilution Western Blot, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunoassay
Formulation 0.2 um filtered solution in PBS, 0.1% BSA
Gene Alias CML
Host Species Mouse
Immunogen CML-KLH
Purification Method Protein G purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 4°C. Do not freeze.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.